Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cercam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233768.2 | XP_006233830.1 | 1 - 2 | 1042 | 102 | |
XM_006233771.2 | XP_006233833.1 | 1 - 2 | 1043 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |