Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapkap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011964.1 | NP_001011964.1 | 4 - 5 | 944 | 82 | |
XM_006233949.2 | XP_006234011.1 | 5 - 6 | 1130 | 82 | ||
XM_006233950.2 | XP_006234012.1 | 4 - 5 | 682 | 82 | ||
XM_008761720.1 | XP_008759942.1 | 4 - 5 | 729 | 82 | ||
XM_008761721.1 | XP_008759943.1 | 3 - 4 | 644 | 82 | ||
XM_008761722.1 | XP_008759944.1 | 2 - 3 | 554 | 82 | ||
GenBank mRNA | BC079073.1 | - | 4 - 5 | 944 | 7122012 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |