Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100770.1 | NP_001094240.1 | 4 - 5 | 455 | 84 | |
XM_006233630.2 | XP_006233692.1 | 4 - 5 | 569 | 84 | ||
XM_006233631.2 | XP_006233693.1 | 3 - 4 | 457 | 84 | ||
GenBank mRNA | BC079372.1 | - | 4 - 5 | 447 | 84 | |
BC158871.1 | - | 4 - 5 | 455 | 7931820 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |