Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tprn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024309.2 | NP_001019480.2 | 3 - 4 | 2202 | 84 | |
GenBank mRNA | BC083928.1 | - | 3 - 4 | 1536 | 7135551 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |