Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spopl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134447.1 | NP_001127919.1 | 2 - 3 | 236 | 114 | |
GenBank mRNA | BC167106.1 | - | 2 - 3 | 236 | 7133920 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |