Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spopl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134447.1 | NP_001127919.1 | 3 - 4 | 368 | 112 | |
GenBank mRNA | BC167106.1 | - | 3 - 4 | 368 | 7133920 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |