Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103351.2 | NP_001096821.1 | 7 - 8 | 1684 | 88 | |
XM_006248411.1 | XP_006248473.1 | 7 - 8 | 1510 | 88 | ||
XM_008768727.1 | XP_008766949.1 | 7 - 8 | 2793 | 88 | ||
XM_008768728.1 | XP_008766950.1 | 6 - 7 | 3438 | 88 | ||
XM_008768729.1 | XP_008766951.1 | 7 - 8 | 1614 | 88 | ||
GenBank mRNA | BC128756.1 | - | 7 - 8 | 1687 | 7105391 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |