Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130424.2 | NP_569108.2 | 4 - 5 | 424 | 67 | |
XM_008768547.1 | XP_008766769.1 | 4 - 5 | 540 | 67 | ||
XM_008768548.1 | XP_008766770.1 | 4 - 5 | 471 | 67 | ||
XM_008768549.1 | XP_008766771.1 | 4 - 5 | 653 | 67 | ||
GenBank mRNA | AB073550.1 | - | 2 - 3 | 322 | 67 | |
BC061712.1 | - | 4 - 5 | 424 | 5598010 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |