Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyttd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY724512.1 | - | 5 - 6 | 753 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |