Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015016.1 | NP_001015016.1 | 5 - 6 | 747 | 65 | |
GenBank mRNA | BC091196.1 | - | 5 - 6 | 747 | 7374499 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |