Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238106.1 | XP_006238168.1 | 4 - 5 | 1371 | 66 | |
GenBank mRNA | BC087577.1 | - | 4 - 5 | 631 | 7307624 | 66 |
FQ220908.1 | - | 4 - 5 | 606 | 66 | ||
FQ230032.1 | - | 4 - 5 | 599 | 66 | ||
M19261.1 | - | 4 - 5 | 598 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |