Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006227646.2 | XP_006227708.1 | 1 - 2 | 156 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |