Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc28a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037789.1 | NP_001032878.1 | 1 - 2 | 96 | 121 | |
XM_006227660.2 | XP_006227722.1 | 1 - 2 | 159 | 121 | ||
GenBank mRNA | BC100268.1 | - | 1 - 2 | 96 | 7385321 | 121 |
FQ212615.1 | - | 1 - 2 | 101 | 121 | ||
FQ213153.1 | - | 1 - 2 | 129 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |