Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lats1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134543.2 | NP_001128015.2 | 5 - 6 | 2940 | 133 | |
XM_006227614.2 | XP_006227676.1 | 4 - 5 | 3582 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Rattus norvegicus Probe spans exons |