Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr2e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113197.1 | NP_001106668.1 | 1 - 2 | 172 | 82 | |
GenBank mRNA | EU316216.1 | - | 1 - 2 | 172 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Nuclear Receptor Rattus norvegicus Probe spans exons |