Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr2e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113197.1 | NP_001106668.1 | 5 - 6 | 735 | 66 | |
GenBank mRNA | EU316216.1 | - | 5 - 6 | 735 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Nuclear Receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |