Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ercc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767061.1 | XP_008765283.1 | 3 - 4 | 547 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |