Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnga3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767042.1 | XP_008765264.1 | 2 - 3 | 466 | 75 | |
GenBank mRNA | AJ272428.1 | - | 2 - 3 | 229 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel Rattus norvegicus Probe spans exons |