Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl21c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106908.1 | NP_001100378.1 | 4 - 5 | 464 | 61 | |
XM_008767022.1 | XP_008765244.1 | 3 - 4 | 1256 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |