Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008758805.1 | XP_008757027.1 | 20 - 21 | 2780 | 52 | |
XM_008758806.1 | XP_008757028.1 | 21 - 22 | 2238 | 52 | ||
XM_008758807.1 | XP_008757029.1 | 20 - 21 | 2034 | 52 | ||
XM_008758808.1 | XP_008757030.1 | 20 - 21 | 2780 | 52 | ||
XM_008758809.1 | XP_008757031.1 | 19 - 20 | 2666 | 52 | ||
XM_008758810.1 | XP_008757032.1 | 19 - 20 | 2589 | 52 | ||
XM_008758811.1 | XP_008757033.1 | 19 - 20 | 2176 | 52 | ||
XM_008758812.1 | XP_008757034.1 | 19 - 20 | 1901 | 52 | ||
XM_008758813.1 | XP_008757035.1 | 19 - 20 | 2165 | 52 | ||
XM_008758814.1 | XP_008757036.1 | 19 - 20 | 4099 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |