Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gkap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012160.1 | NP_001012160.1 | 5 - 6 | 765 | 138 | |
XM_006253559.2 | XP_006253621.1 | 5 - 6 | 620 | 138 | ||
XM_006253561.2 | XP_006253623.1 | 4 - 5 | 542 | 138 | ||
GenBank mRNA | BC083717.1 | - | 5 - 6 | 765 | 7190699 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |