Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001056825.4 | XP_001056825.3 | 8 - 9 | 1358 | 114 | |
XM_006253546.2 | XP_006253608.1 | 8 - 9 | 1638 | 114 | ||
XM_006253547.2 | XP_006253609.1 | 9 - 10 | 1600 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |