Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfbi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053802.1 | NP_446254.1 | 11 - 12 | 1573 | 123 | |
GenBank mRNA | AF305713.1 | - | 3 - 4 | 341 | 123 | |
CA340798.1 | - | 4 - 5 | 547 | 123 | ||
FQ229143.1 | - | 2 - 3 | 160 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |