Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011966.1 | NP_001011966.1 | 11 - 12 | 1266 | 92 | |
XM_006235883.2 | XP_006235945.1 | 10 - 11 | 1363 | 92 | ||
XM_006235884.2 | XP_006235946.1 | 10 - 11 | 1363 | 92 | ||
XM_006235885.2 | XP_006235947.1 | 10 - 11 | 1327 | 92 | ||
XM_006235886.2 | XP_006235948.1 | 11 - 12 | 1350 | 92 | ||
XM_006235887.2 | XP_006235949.1 | 11 - 12 | 1314 | 92 | ||
XM_006235888.2 | XP_006235950.1 | 10 - 11 | 1363 | 92 | ||
XM_006235889.2 | XP_006235951.1 | 10 - 11 | 1327 | 92 | ||
GenBank mRNA | BC085349.1 | - | 11 - 12 | 1266 | 7107899 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |