Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191754.1 | NP_001178683.1 | 1 | 39 | 117 | |
XM_008763476.1 | XP_008761698.1 | 1 | 364 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon |