Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gatsl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025128.1 | NP_001020299.1 | 8 - 9 | 980 | 71 | |
XM_006251335.2 | XP_006251397.1 | 8 - 9 | 1048 | 71 | ||
XM_006251336.2 | XP_006251398.1 | 8 - 9 | 1048 | 71 | ||
GenBank mRNA | BC091274.1 | - | 8 - 9 | 980 | 7383249 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |