Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106009.1 | NP_001099479.1 | 2 - 3 | 463 | 141 | |
XM_008770199.1 | XP_008768421.1 | 2 - 3 | 464 | 141 | ||
XM_008770200.1 | XP_008768422.1 | 2 - 3 | 464 | 141 | ||
XM_008770201.1 | XP_008768423.1 | 2 - 3 | 464 | 141 | ||
XM_008770202.1 | XP_008768424.1 | 2 - 3 | 464 | 141 | ||
XM_008770203.1 | XP_008768425.1 | 2 - 3 | 464 | 141 | ||
GenBank mRNA | BC168881.1 | - | 2 - 3 | 348 | 8363537 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |