Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130988.1 | NP_001124460.1 | 3 - 4 | 323 | 150 | |
GenBank mRNA | BC167755.1 | - | 3 - 4 | 323 | 8363815 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |