Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec14l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022608.1 | NP_072130.1 | 4 - 5 | 258 | 70 | |
GenBank mRNA | AJ132352.1 | - | 4 - 5 | 273 | 70 | |
BC162038.1 | - | 4 - 5 | 258 | 7099625 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |