Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191680.1 | NP_001178609.1 | 9 - 10 | 1122 | 111 | |
XM_006251243.2 | XP_006251305.1 | 10 - 11 | 1424 | 111 | ||
XM_006251244.2 | XP_006251306.1 | 10 - 11 | 1421 | 111 | ||
XM_006251245.2 | XP_006251307.1 | 10 - 11 | 1257 | 111 | ||
XM_006251246.2 | XP_006251308.1 | 10 - 11 | 1254 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |