Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021739.2 | NP_068507.2 | 14 - 15 | 1147 | 88 | |
XM_006251170.2 | XP_006251232.1 | 14 - 15 | 1246 | 88 | ||
XM_006251171.2 | XP_006251233.1 | 14 - 15 | 1246 | 88 | ||
XM_006251176.2 | XP_006251238.1 | 14 - 15 | 1227 | 88 | ||
GenBank mRNA | M16112.1 | - | 14 - 15 | 1127 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Protein Phosphatase Detect Genomic DNA Rattus norvegicus Probe spans exons |