Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181365.2 | NP_852030.1 | 918 | 144 | ||
GenBank mRNA | AF345444.1 | - | 918 | 144 | ||
AF345445.1 | - | 4 - 5 | 488 | 144 | ||
AF453245.1 | - | 5 - 6 | 541 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |