Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012032.1 | NP_001012032.1 | 5 - 6 | 919 | 110 | |
XM_006250652.2 | XP_006250714.1 | 6 - 7 | 1125 | 110 | ||
XM_006250654.2 | XP_006250716.1 | 6 - 7 | 3707 | 110 | ||
XM_008770017.1 | XP_008768239.1 | 6 - 7 | 1786 | 110 | ||
XM_008770018.1 | XP_008768240.1 | 5 - 6 | 1638 | 110 | ||
GenBank mRNA | AB108670.1 | - | 5 - 6 | 1121 | 110 | |
BC085797.1 | - | 5 - 6 | 919 | 7109913 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |