Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam175a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009633.1 | NP_001009633.1 | 5 - 6 | 525 | 75 | |
GenBank mRNA | BC088408.1 | - | 5 - 6 | 525 | 7131771 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |