Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105995.1 | NP_001099465.1 | 2 - 3 | 506 | 57 | |
XM_008770004.1 | XP_008768226.1 | 4 - 5 | 841 | 57 | ||
XM_008770005.1 | XP_008768227.1 | 4 - 5 | 745 | 57 | ||
XM_008770006.1 | XP_008768228.1 | 4 - 5 | 683 | 57 | ||
XM_008770007.1 | XP_008768229.1 | 3 - 4 | 510 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |