Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016996.1 | NP_058692.1 | 2 - 3 | 764 | 105 | |
GenBank mRNA | AY214122.1 | - | 2 - 3 | 754 | 105 | |
U10354.1 | - | 2 - 3 | 764 | 105 | ||
U20289.1 | - | 1 - 2 | 390 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Rattus norvegicus Probe spans exons |