Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134350.2 | NP_599177.2 | 11 - 12 | 1545 | 119 | |
XM_006248150.2 | XP_006248212.1 | 13 - 14 | 1689 | 119 | ||
XM_008768568.1 | XP_008766790.1 | 13 - 14 | 1623 | 119 | ||
GenBank mRNA | BC099808.1 | - | 11 - 12 | 1545 | 7938638 | 119 |
X52712.1 | - | 11 - 12 | 1620 | 119 | ||
X52713.1 | - | 10 - 11 | 1425 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |