Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134350.2 | NP_599177.2 | 4 - 5 | 550 | 76 | |
XM_006248150.2 | XP_006248212.1 | 6 - 7 | 694 | 76 | ||
XM_008768568.1 | XP_008766790.1 | 6 - 7 | 628 | 76 | ||
GenBank mRNA | BC099808.1 | - | 4 - 5 | 550 | 7938638 | 76 |
X52712.1 | - | 4 - 5 | 625 | 76 | ||
X52713.1 | - | 3 - 4 | 430 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |