Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022265.2 | NP_071601.2 | 7 - 8 | 992 | 77 | |
XM_006231613.2 | XP_006231675.1 | 8 - 9 | 1273 | 77 | ||
XM_006231614.2 | XP_006231676.1 | 8 - 9 | 1325 | 77 | ||
XM_006231615.2 | XP_006231677.1 | 8 - 9 | 1171 | 77 | ||
XM_006231616.2 | XP_006231678.1 | 8 - 9 | 1148 | 77 | ||
GenBank mRNA | AF239739.2 | - | 7 - 8 | 992 | 77 | |
BC167751.1 | - | 8 - 9 | 1180 | 9027822 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Rattus norvegicus Probe spans exons |