Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017075.2 | NP_058771.2 | 10 - 11 | 1076 | 89 | |
GenBank mRNA | D00512.1 | - | 10 - 11 | 1008 | 89 | |
D13921.1 | - | 10 - 11 | 1008 | 89 | ||
FM098848.1 | - | 5 - 6 | 458 | 89 | ||
FQ228108.1 | - | 10 - 11 | 1062 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |