Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109437.2 | NP_001102907.1 | 2 - 3 | 429 | 111 | |
GenBank mRNA | BC166797.1 | - | 2 - 3 | 429 | 7938214 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |