Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hemk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106853.1 | NP_001100323.1 | 3 - 4 | 514 | 65 | |
XM_006243728.2 | XP_006243790.1 | 4 - 5 | 804 | 65 | ||
XM_006243729.2 | XP_006243791.1 | 4 - 5 | 1124 | 65 | ||
XM_006243730.2 | XP_006243792.1 | 3 - 4 | 879 | 65 | ||
XM_008766502.1 | XP_008764724.1 | 4 - 5 | 739 | 65 | ||
GenBank mRNA | BC161944.1 | - | 3 - 4 | 508 | 9024451 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |