Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133427.3 | NP_596918.3 | 8 - 9 | 796 | 97 | |
XM_006243472.2 | XP_006243534.1 | 7 - 8 | 782 | 97 | ||
XM_008766437.1 | XP_008764659.1 | 8 - 9 | 955 | 97 | ||
GenBank mRNA | AF307840.1 | - | 7 - 8 | 555 | 97 | |
AY321370.1 | - | 7 - 8 | 555 | 97 | ||
AY321371.1 | - | 7 - 8 | 555 | 97 | ||
BC080240.1 | - | 8 - 9 | 774 | 7099950 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |