Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF157498.1 | - | 5 - 6 | 837 | 75 | |
BC087694.1 | - | 3 - 4 | 495 | 75 | ||
D28111.1 | - | 4 - 5 | 621 | 75 | ||
X87900.1 | - | 4 - 5 | 534 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |