Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244005.1 | XP_006244067.1 | 5 - 6 | 834 | 64 | |
XM_006244006.2 | XP_006244068.1 | 5 - 6 | 1490 | 64 | ||
XM_006244007.1 | XP_006244069.1 | 5 - 6 | 648 | 64 | ||
XM_236647.7 | XP_236647.6 | 5 - 6 | 834 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |