Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amotl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031717.1 | NP_113905.1 | 8 - 9 | 2240 | 92 | |
XM_006243680.2 | XP_006243742.1 | 8 - 9 | 2342 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |