Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glyctk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109449.2 | NP_001102919.1 | 2 - 3 | 465 | 97 | |
XM_006243856.2 | XP_006243918.1 | 2 - 3 | 740 | 97 | ||
XM_006243857.2 | XP_006243919.1 | 2 - 3 | 523 | 97 | ||
GenBank mRNA | BC105621.1 | - | 2 - 3 | 465 | 7454021 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |