Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tusc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109297.1 | NP_001102767.1 | 2 - 3 | 809 | 107 | |
GenBank mRNA | BC082798.1 | - | 2 - 3 | 293 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |