Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271306.1 | NP_001258235.1 | 452 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |