Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nprl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025744.2 | NP_001020915.2 | 6 - 7 | 815 | 73 | |
GenBank mRNA | DV714395.1 | - | 2 - 3 | 223 | 73 | |
FQ209669.1 | - | 5 - 6 | 888 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |